Assay Details
Target Gene Details
Entrez Gene ID: | 73167 |
Gene Name: | Rho GTPase activating protein 8 |
Gene Aliases: |
3110043J09Rik |
Location: |
Chr.15:84720052-84772857 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap8 | NM_001164627.1 | 3 | 132 | NP_001158099.1 |
| NM_001164628.1 | 2 | 124 | NP_001158100.1 | |
| NM_001205334.1 | 3 | 152 | NP_001192263.1 | |
| NM_028455.4 | 2 | 144 | NP_082731.2 | |
| AK014171.1 | 2 | 95 | BAB29190.1 | |
| AK040142.1 | 2 | 94 | ||
| AK086816.1 | 2 | 98 | BAC39750.1 | |
| AK088242.1 | 2 | 100 | BAC40230.1 | |
| BC005563.1 | 1 | 87 | AAH05563.1 | |
| BC010306.1 | 2 | 114 | AAH10306.2 | |
| BY723460.1 | 2 | 118 |
More Information
Set Membership: |
|

Genomic Map