Assay Details
Target Gene Details
Entrez Gene ID: | 328572 |
Gene Name: | E1A binding protein p300 |
Gene Aliases: |
A430090G16, A730011L11, KAT3B, p300, p300 HAT |
Location: |
Chr.15:81586214-81652077 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ep300 | NM_177821.6 | NP_808489.4 | ||
| AI049018.1 | ||||
| AK139715.1 | ||||
| BC144976.1 | ||||
| BC150681.1 | ||||
| CN708842.1 |
More Information
Set Membership: |
|

Genomic Map