Assay Details
Target Gene Details
Entrez Gene ID: | 14114 |
Gene Name: | fibulin 1 |
Gene Aliases: |
- |
Location: |
Chr.15:85205944-85286294 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbln1 | NM_010180.2 | 9 | 1120 | NP_034310.2 |
AK035388.1 | 9 | 1073 | BAC29054.1 | |
AK083573.1 | 9 | 1123 | ||
AK085989.1 | 9 | 1063 | ||
AK086451.1 | 9 | 1121 | BAC39669.1 | |
AK137511.1 | 9 | 1063 | ||
AK159646.1 | 9 | 1075 | BAE35258.1 | |
AK188438.1 | 2 | 101 | ||
BC007140.1 | 9 | 1064 | AAH07140.1 | |
EU543224.1 | 9 | 1058 | ||
X70854.1 | 9 | 1117 | CAA50207.1 |
