Assay Details
Target Gene Details
Entrez Gene ID: | 56149 |
Gene Name: | GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein |
Gene Aliases: |
tamalin |
Location: |
Chr.15:101224207-101232756 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grasp | NM_019518.3 | NP_062391.3 | ||
| AB041603.1 | BAA95086.1 | |||
| AF236099.1 | AAF36997.1 | |||
| BC046307.1 | AAH46307.1 | |||
| BY730445.1 |
More Information
Set Membership: |
|

Genomic Map