Assay Details
Target Gene Details
Entrez Gene ID: | 11564 |
Gene Name: | adenylosuccinate lyase |
Gene Aliases: |
Adl, Asl |
Location: |
Chr.15:80948490-80970947 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adsl | NM_009634.6 | 12 | 1375 | NP_033764.2 |
| AK049372.1 | 12 | 1347 | BAC33717.1 | |
| AK051111.1 | 12 | 1347 | BAC34529.1 | |
| AK152092.1 | 12 | 1347 | BAE30940.1 | |
| AK166019.1 | 12 | 1343 | BAE38524.1 | |
| AK168906.1 | 12 | 1340 | BAE40720.1 | |
| AK170216.1 | 10 | 1410 | ||
| AK171725.1 | 12 | 1356 | BAE42633.1 | |
| BC020187.1 | 12 | 1351 | AAH20187.1 | |
| U20225.1 | 12 | 1351 | AAB60684.1 |
More Information
Set Membership: |
|

Genomic Map