Assay Details
Target Gene Details
Entrez Gene ID: | 77627 |
Gene Name: | EF-hand calcium binding domain 6 |
Gene Aliases: |
4931407K02Rik, 4932408N08Rik, bM150J22.2, mKIAA1672 |
Location: |
Chr.15:83866712-84075025 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab6 | NM_001161628.1 | 9 | 1350 | NP_001155100.1 |
| NM_001161629.1 | 7 | 838 | NP_001155101.1 | |
| NM_029946.4 | 26 | 3457 | NP_084222.4 | |
| AK016516.1 | 7 | 943 | ||
| AK019850.1 | 9 | 1350 | BAB31880.1 | |
| AK163363.1 | 4 | 528 | ||
| AK220412.1 | 9 | 1269 | BAD90458.1 | |
| BC056955.1 | 14 | 1860 | AAH56955.1 | |
| BC065111.1 | 26 | 3457 | AAH65111.1 |
More Information
Set Membership: |
|

Genomic Map