Assay Details
Target Gene Details
Entrez Gene ID: | 14663 |
Gene Name: | glycosylation dependent cell adhesion molecule 1 |
Gene Aliases: |
MC26, Sgp50, glyCAM-1 |
Location: |
Chr.15:103562759-103565081 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glycam1 | NM_001289587.1 | NP_001276516.1 | ||
NM_008134.3 | NP_032160.1 | |||
AK021358.1 | BAB32385.1 |
