Assay Details
Target Gene Details
Entrez Gene ID: | 11564 |
Gene Name: | adenylosuccinate lyase |
Gene Aliases: |
Adl, Asl |
Location: |
Chr.15:80948490-80970947 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adsl | NM_009634.6 | NP_033764.2 | ||
| AK049372.1 | BAC33717.1 | |||
| AK051111.1 | BAC34529.1 | |||
| AK152092.1 | BAE30940.1 | |||
| AK166019.1 | BAE38524.1 | |||
| AK168906.1 | BAE40720.1 | |||
| AK170216.1 | ||||
| AK171725.1 | BAE42633.1 | |||
| AK190788.1 | ||||
| BC020187.1 | AAH20187.1 | |||
| U20225.1 | AAB60684.1 |
More Information
Set Membership: |
|

Genomic Map