Assay Details
Target Gene Details
Entrez Gene ID: | 239420 |
Gene Name: | CUB and Sushi multiple domains 3 |
Gene Aliases: |
4930500N14Rik, mKIAA1894 |
Location: |
Chr.15:47580633-48792231 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 63 - Exon 64 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csmd3 | NM_001081391.2 | NP_001074860.2 | ||
AK078076.1 | ||||
AK122567.1 | BAC65849.1 | |||
AK162216.1 | BAE36794.1 |
