Assay Details
Target Gene Details
Entrez Gene ID: | 13196 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain1 |
Gene Aliases: |
AV239055, DEF-1, Ddef1, PAP, mKIAA1249, s19 |
Location: |
Chr.15:64086840-64382919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asap1 | NM_001276461.1 | NP_001263390.1 | ||
| NM_001276462.1 | NP_001263391.1 | |||
| NM_001276463.1 | NP_001263392.1 | |||
| NM_001276467.1 | NP_001263396.1 | |||
| NM_010026.3 | NP_034156.2 | |||
| AF075461.1 | AAC98349.1 | |||
| AF075462.1 | AAC98350.1 | |||
| AK122477.1 | BAC65759.1 | |||
| AK146545.1 | BAE27250.1 | |||
| AK147689.1 | ||||
| BC048818.1 | AAH48818.1 | |||
| BC094581.1 | AAH94581.1 | |||
| CB527210.1 | ||||
| CD355387.1 | ||||
| CN455356.1 |

Genomic Map