Assay Details
Target Gene Details
Entrez Gene ID: | 109075 |
Gene Name: | exosome component 4 |
Gene Aliases: |
1110039I09Rik, 1500001N04Rik, Rrp41 |
Location: |
Chr.15:76327397-76330670 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exosc4 | NM_175399.4 | 2 | 327 | NP_780608.1 |
| AK005101.2 | 2 | 264 | ||
| AK083308.1 | 2 | 286 | ||
| AK089890.1 | 2 | 328 | BAC40987.1 | |
| AK144062.1 | 2 | 242 | BAE25679.1 | |
| AK167629.1 | 2 | 290 | BAE39680.1 | |
| BC012277.1 | 2 | 272 | AAH12277.1 |
Target Gene Details
Entrez Gene ID: | 75475 |
Gene Name: | 5-oxoprolinase (ATP-hydrolysing) |
Gene Aliases: |
1700010G02Rik |
Location: |
Chr.15:76296603-76331101 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oplah |
More Information
Set Membership: |
|

Genomic Map