Assay Details
Target Gene Details
Entrez Gene ID: | 16185 |
Gene Name: | interleukin 2 receptor, beta chain |
Gene Aliases: |
CD122, IL-15Rbeta, IL15Rbeta, Il-2/15Rbeta, Il-2Rbeta, p70 |
Location: |
Chr.15:78479256-78511627 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il2rb | NM_008368.4 | NP_032394.1 | ||
| AK017288.1 | BAB30674.1 | |||
| AK157572.1 | BAE34125.1 | |||
| AK169481.1 | BAE41196.1 | |||
| BC026869.1 | AAH26869.1 |
More Information
Set Membership: |
|

Genomic Map