Assay Details
Target Gene Details
Entrez Gene ID: | 16185 |
Gene Name: | interleukin 2 receptor, beta chain |
Gene Aliases: |
CD122, IL-15Rbeta, IL15Rbeta, Il-2/15Rbeta, Il-2Rbeta, p70 |
Location: |
Chr.15:78479256-78511627 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il2rb | NM_008368.4 | 6 | 572 | NP_032394.1 |
| AK017288.1 | 6 | 572 | BAB30674.1 | |
| AK035289.1 | 6 | 525 | BAC29017.1 | |
| AK157572.1 | 6 | 573 | BAE34125.1 | |
| AK169481.1 | 6 | 553 | BAE41196.1 | |
| BC026869.1 | 6 | 545 | AAH26869.1 | |
| BC099616.1 | 6 | 546 | AAH99616.1 | |
| M28052.1 | 6 | 529 | AAA39283.1 |
More Information
Set Membership: |
|

Genomic Map