Assay Details
Target Gene Details
Entrez Gene ID: | 67040 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 |
Gene Aliases: |
2610007K22Rik, A430025E01Rik, AI047725, C80929, Gm926, p72 |
Location: |
Chr.15:79527696-79546764 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx17 | NM_001040187.1 | 8 | 1431 | NP_001035277.1 |
NM_199079.1 | 8 | 1250 | NP_951061.1 | |
NM_199080.2 | 8 | 1431 | NP_951062.1 | |
AK039888.1 | 8 | 1250 | BAC30474.1 | |
AK151061.1 | 8 | 1206 | BAE30078.1 | |
AK152749.1 | 8 | 1207 | ||
AK152831.1 | 8 | 1206 | BAE31531.1 | |
AK161001.1 | 8 | 1247 | ||
AK161012.1 | 8 | 1246 | BAE36146.1 | |
BC062910.1 | 8 | 1236 | AAH62910.1 | |
BC096036.1 | 8 | 1431 | AAH96036.1 | |
BI655302.1 | 1 | 103 |
