Assay Details
Target Gene Details
Entrez Gene ID: | 268780 |
Gene Name: | EGF-like, fibronectin type III and laminin G domains |
Gene Aliases: |
5930412K08, AU040377 |
Location: |
Chr.15:7206120-7398746 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Egflam | NM_001289496.1 | NP_001276425.1 | ||
| NM_001289498.1 | NP_001276427.1 | |||
| NM_178748.6 | NP_848863.1 | |||
| AK031136.1 | BAC27271.1 | |||
| AK033332.1 | BAC28235.1 | |||
| AK037223.1 | BAC29762.1 | |||
| AK041546.1 | BAC30982.1 | |||
| AK158909.1 | ||||
| BC051455.1 | AAH51455.1 | |||
| BC095994.1 | AAH95994.1 | |||
| BC150710.1 | ||||
| DQ223720.1 |
More Information
Set Membership: |
|

Genomic Map