Assay Details
Target Gene Details
Entrez Gene ID: | 66120 |
Gene Name: | FK506 binding protein 11 |
Gene Aliases: |
1110002O23Rik, FKBP-11 |
Location: |
Chr.15:98724366-98728198 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp11 | NM_024169.3 | 6 | 659 | NP_077131.2 |
AK003331.1 | 6 | 660 | BAB22719.1 | |
AK007813.1 | 6 | 623 | ||
AK019132.1 | 6 | 601 | BAB31559.1 | |
BC037596.1 | 6 | 615 | AAH37596.1 |
