Assay Details
Target Gene Details
Entrez Gene ID: | 74351 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 23 |
Gene Aliases: |
3110082M05Rik, 4921506D17Rik |
Location: |
Chr.15:98645379-98662936 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx23 | NM_001080981.1 | NP_001074450.1 |
