Assay Details
Target Gene Details
Entrez Gene ID: | 14106 |
Gene Name: | forkhead box H1 |
Gene Aliases: |
Fast1, Fast2, fast-1, fast-2 |
Location: |
Chr.15:76667829-76670317 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxh1 | NM_007989.4 | NP_032015.1 | ||
AB221574.1 | ||||
AF069303.1 | AAC79808.1 | |||
AF079514.1 | AAD12486.1 | |||
AF110506.1 | AAD14683.1 | |||
AF177770.1 | AAD55949.1 | |||
BC119156.1 | ||||
BC137781.1 |
