Assay Details
Target Gene Details
Entrez Gene ID: | 320865 |
Gene Name: | cadherin 18 |
Gene Aliases: |
B230220E17Rik, Cdh14l |
Location: |
Chr.15:22548899-23474432 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh18 | NM_001081299.1 | NP_001074768.1 | ||
| AK045672.1 | BAC32451.1 | |||
| AK137430.1 |
More Information
Set Membership: |
|

Genomic Map