Assay Details
Target Gene Details
Entrez Gene ID: | 12563 |
Gene Name: | cadherin 6 |
Gene Aliases: |
K-cadherin, cad6, cadherin-6 |
Location: |
Chr.15:13028699-13173689 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh6 | NM_007666.3 | 6 | 1161 | NP_031692.2 |
| BC107008.1 | 5 | 902 | AAI07009.1 | |
| BC107013.1 | 5 | 902 | AAI07014.1 | |
| D82029.1 | 5 | 902 | BAA11516.1 |
More Information
Set Membership: |
|

Genomic Map