Assay Details
Target Gene Details
Entrez Gene ID: | 69440 |
Gene Name: | DENN/MADD domain containing 6B |
Gene Aliases: |
1700027J05Rik, AI414881, Fam116b |
Location: |
Chr.15:89182213-89196474 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd6b | NM_027081.3 | 17 | 1476 | NP_081357.2 |
| AK006427.1 | 8 | 613 | BAB24583.1 | |
| AK084555.1 | 15 | 1502 | ||
| AK154949.1 | 14 | 2056 | ||
| BC139003.1 | 17 | 1450 | ||
| BQ176037.1 | 4 | 552 |

Genomic Map