Assay Details
Target Gene Details
Entrez Gene ID: | 268816 |
Gene Name: | maestro heat-like repeat family member 5 |
Gene Aliases: |
Gm628 |
Location: |
Chr.15:73786936-73839671 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mroh5 | NM_001033365.2 | NP_001028537.1 | ||
| AK158799.1 | BAE34672.1 | |||
| BC150895.1 | ||||
| BC172046.1 |

Genomic Map