Assay Details
Target Gene Details
Entrez Gene ID: | 16581 |
Gene Name: | kinesin family member C2 |
Gene Aliases: |
- |
Location: |
Chr.15:76659837-76668196 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kifc2 | NM_010630.2 | NP_034760.2 | ||
| AK144304.1 | BAE25823.1 | |||
| D49545.1 | BAA19677.1 | |||
| U92949.1 | AAB51397.1 |
More Information
Set Membership: |
|

Genomic Map