Assay Details
Target Gene Details
Entrez Gene ID: | 21847 |
Gene Name: | Kruppel-like factor 10 |
Gene Aliases: |
AI115143, EGR[a], Egral, Gdnfif, TIEG-1, Tieg, Tieg1, mGIF |
Location: |
Chr.15:38291464-38300711 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf10 | NM_001289471.1 | NP_001276400.1 | ||
| NM_013692.3 | NP_038720.1 | |||
| AF049879.1 | AAC77796.1 | |||
| AF064088.1 | AAC16734.1 | |||
| AK004906.1 | ||||
| AK043433.1 | BAC31547.1 | |||
| AK079679.1 | ||||
| AK154719.1 | BAE32785.1 | |||
| BC003316.1 | AAH03316.1 | |||
| Z36270.1 | CAA85283.1 |
More Information
Set Membership: |
|

Genomic Map