Assay Details
Target Gene Details
Entrez Gene ID: | 69440 |
Gene Name: | DENN/MADD domain containing 6B |
Gene Aliases: |
1700027J05Rik, AI414881, Fam116b |
Location: |
Chr.15:89182213-89196474 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd6b | NM_027081.3 | NP_081357.2 | ||
| AK084555.1 | ||||
| AK154949.1 | ||||
| BC139003.1 | ||||
| BQ444357.1 |

Genomic Map