Assay Details
Target Gene Details
Entrez Gene ID: | 73225 |
Gene Name: | family with sequence similarity 118, member A |
Gene Aliases: |
- |
Location: |
Chr.15:85037015-85116933 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam118a | NM_133750.4 | NP_598511.1 | ||
| AK082151.1 | BAC38423.1 | |||
| AK143690.1 | BAE25500.1 | |||
| AK180870.1 | ||||
| AK189276.1 | ||||
| BC016425.1 | AAH16425.1 | |||
| BC066174.1 | AAH66174.1 | |||
| CJ084461.1 |

Genomic Map