Assay Details
Target Gene Details
Entrez Gene ID: | 170787 |
Gene Name: | histone deacetylase 10 |
Gene Aliases: |
AW548891, Hd10 |
Location: |
Chr.15:89123303-89128700 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac10 | NM_199198.2 | NP_954668.2 | ||
NR_028447.1 | ||||
NR_028448.1 | ||||
NR_028449.1 | ||||
AK040235.1 | ||||
AK045028.1 | ||||
AK154045.1 | BAE32336.1 | |||
BC013700.1 | ||||
BC064018.1 | AAH64018.1 | |||
BU706947.1 |
