Assay Details
Target Gene Details
Entrez Gene ID: | 223691 |
Gene Name: | eukaryotic translation initiation factor 3, subunit L |
Gene Aliases: |
0610011H21Rik, D15N1e, Eif3eip, Eif3ip, Eif3s6ip, HSP-66Y, PAF67 |
Location: |
Chr.15:79075223-79094400 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3l | NM_145139.2 | NP_660121.2 | ||
| AJ310346.1 | CAC84554.1 | |||
| AK032936.1 | BAC28090.1 | |||
| AK075584.1 | BAC35837.1 | |||
| AK150895.1 | BAE29941.1 | |||
| AK151175.1 | BAE30177.1 | |||
| AK167640.1 | BAE39691.1 | |||
| AK168009.1 | BAE39996.1 | |||
| AK168295.1 | BAE40238.1 | |||
| BC024463.1 | AAH24463.1 |
More Information
Set Membership: |
|

Genomic Map