Assay Details
Target Gene Details
Entrez Gene ID: | 170787 |
Gene Name: | histone deacetylase 10 |
Gene Aliases: |
AW548891, Hd10 |
Location: |
Chr.15:89123303-89128700 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac10 | NM_199198.2 | 7 | 884 | NP_954668.2 |
NR_028447.1 | 7 | 884 | ||
NR_028448.1 | 6 | 815 | ||
NR_028449.1 | 7 | 884 | ||
AK040235.1 | 7 | 836 | ||
AK045028.1 | 6 | 1095 | ||
AK154045.1 | 6 | 779 | BAE32336.1 | |
BC013700.1 | 7 | 837 | ||
BC064018.1 | 7 | 811 | AAH64018.1 | |
BU706947.1 | 7 | 625 |
