Assay Details
Target Gene Details
Entrez Gene ID: | 12614 |
Gene Name: | cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila) |
Gene Aliases: |
Adgrc1, Crsh, Scy, crash |
Location: |
Chr.15:85898758-86034217 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Celsr1 | NM_009886.2 | 33 | 8718 | NP_034016.2 |
| AF031572.1 | 33 | 8718 | AAC68836.1 | |
| AK220170.1 | 32 | 4609 | BAD90355.1 | |
| CV561900.1 | 5 | 490 |
More Information
Set Membership: |
|

Genomic Map