Assay Details
Target Gene Details
Entrez Gene ID: | 246277 |
Gene Name: | cysteine sulfinic acid decarboxylase |
Gene Aliases: |
Csd |
Location: |
Chr.15:102176997-102204231 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csad | NM_144942.4 | NP_659191.1 | ||
| AK005015.1 | BAB23747.1 | |||
| AY033912.1 | AAK60398.1 |
More Information
Set Membership: |
|

Genomic Map