Assay Details
Target Gene Details
Entrez Gene ID: | 75705 |
Gene Name: | eukaryotic translation initiation factor 4B |
Gene Aliases: |
2310046H11Rik, AL024095, C85189, Eif4a2 |
Location: |
Chr.15:102073773-102097173 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4b | NM_145625.3 | NP_663600.2 | ||
| AK045250.1 | BAC32280.1 | |||
| AK077423.1 | BAC36793.1 | |||
| AK148012.1 | BAE28289.1 | |||
| AK161693.1 | BAE36536.1 | |||
| AK163266.1 | ||||
| AK166363.1 | BAE38733.1 | |||
| AK168350.1 | BAE40287.1 | |||
| AK168425.1 | ||||
| AK170256.1 | BAE41664.1 | |||
| AK170354.1 | BAE41740.1 | |||
| AK177300.1 | ||||
| AK180065.1 | ||||
| AK180816.1 | ||||
| AK186733.1 | ||||
| AK192567.1 | ||||
| AK195122.1 | ||||
| AK198498.1 | ||||
| AK203780.1 | ||||
| AK205492.1 | ||||
| AK207103.1 | ||||
| AK207417.1 | ||||
| AK210570.1 | ||||
| AK212755.1 | ||||
| AK212975.1 | ||||
| AK213592.1 | ||||
| AK214822.1 | ||||
| AK218019.1 | ||||
| BC007171.1 | AAH07171.1 | |||
| BC147746.1 | ||||
| BC147748.1 |
More Information
Set Membership: |
|

Genomic Map