Assay Details
Target Gene Details
Entrez Gene ID: | 14731 |
Gene Name: | GPI anchor attachment protein 1 |
Gene Aliases: |
C80044, mGAA1 |
Location: |
Chr.15:76331294-76334900 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpaa1 | NM_010331.2 | 10 | 1490 | NP_034461.1 |
AB002136.1 | 10 | 1419 | BAA82589.1 | |
AB006970.1 | 10 | 1426 | BAB03274.1 | |
AK077518.1 | 10 | 1490 | BAC36840.1 | |
AK164219.1 | 10 | 1436 | BAE37686.1 | |
AK164978.1 | 10 | 1490 | BAE37988.1 | |
AK168057.1 | 10 | 1693 | ||
AK177221.1 | ||||
BC006697.1 | 10 | 1429 | AAH06697.1 |
