Assay Details
Target Gene Details
Entrez Gene ID: | 14731 |
Gene Name: | GPI anchor attachment protein 1 |
Gene Aliases: |
C80044, mGAA1 |
Location: |
Chr.15:76331294-76334900 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpaa1 | NM_010331.2 | 11 | 1589 | NP_034461.1 |
| AB002136.1 | 11 | 1518 | BAA82589.1 | |
| AB006970.1 | 11 | 1525 | BAB03274.1 | |
| AK077518.1 | 11 | 1589 | BAC36840.1 | |
| AK164219.1 | 11 | 1535 | BAE37686.1 | |
| AK164978.1 | 11 | 1589 | BAE37988.1 | |
| AK168057.1 | 11 | 1792 | ||
| BC006697.1 | 11 | 1528 | AAH06697.1 |

Genomic Map