Assay Details
Target Gene Details
Entrez Gene ID: | 110308 |
Gene Name: | keratin 5 |
Gene Aliases: |
3300001P10Rik, AW146334, CK5, K5, Krt2-5, Tfip8 |
Location: |
Chr.15:101707070-101712891 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt5 | NM_027011.2 | NP_081287.1 | ||
| AK014363.1 | BAB29298.2 | |||
| BC006780.1 | AAH06780.1 | |||
| BC108361.1 |
More Information
Set Membership: |
|

Genomic Map