Assay Details
Target Gene Details
Entrez Gene ID: | 105859 |
Gene Name: | cold shock domain containing C2, RNA binding |
Gene Aliases: |
AI415250, AI481750, Pippin |
Location: |
Chr.15:81936759-81950941 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csdc2 | NM_145473.3 | NP_663448.2 | ||
| AK028916.1 | BAC26194.1 | |||
| AK132493.1 | BAE21201.1 | |||
| BB871178.1 | ||||
| BC016109.1 | AAH16109.1 |
More Information
Set Membership: |
|

Genomic Map