Assay Details
Target Gene Details
Entrez Gene ID: | 26912 |
Gene Name: | glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase) |
Gene Aliases: |
AI526977, Kbl |
Location: |
Chr.15:79030874-79042531 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcat | NM_001161712.1 | NP_001155184.1 | ||
| NM_013847.4 | NP_038875.3 | |||
| AF093403.1 | AAC99774.1 | |||
| AK007395.1 | ||||
| AK013138.1 | BAB28673.1 | |||
| AK075782.1 | ||||
| AK076469.1 | ||||
| AK087433.1 | BAC39872.1 | |||
| BC024107.1 | AAH24107.1 |
More Information
Set Membership: |
|

Genomic Map