Assay Details
Target Gene Details
Entrez Gene ID: | 14731 |
Gene Name: | GPI anchor attachment protein 1 |
Gene Aliases: |
C80044, mGAA1 |
Location: |
Chr.15:76331294-76334900 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpaa1 | NM_010331.2 | NP_034461.1 | ||
AB002136.1 | BAA82589.1 | |||
AB006970.1 | BAB03274.1 | |||
AK077518.1 | BAC36840.1 | |||
AK164219.1 | BAE37686.1 | |||
AK164978.1 | BAE37988.1 | |||
AK168057.1 | 2 | 376 | ||
BC006697.1 | AAH06697.1 |
