Assay Details
Target Gene Details
Entrez Gene ID: | 223920 |
Gene Name: | sterol O-acyltransferase 2 |
Gene Aliases: |
ACAT2, D15Wsu97e |
Location: |
Chr.15:102150414-102163436 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Soat2 | NM_146064.1 | NP_666176.1 | ||
| AF078751.1 | AAC64057.1 | |||
| BC025931.1 | AAH25931.1 |
More Information
Set Membership: |
|

Genomic Map