Assay Details
Target Gene Details
Entrez Gene ID: | 20356 |
Gene Name: | sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A |
Gene Aliases: |
5930434A13, 9130201M22Rik, AI464145, Semaf, semF |
Location: |
Chr.15:32244742-32696341 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema5a | NM_009154.2 | 22 | 3697 | NP_033180.2 |
| AK077021.1 | 13 | 2368 | BAC36572.1 | |
| AK141612.1 | 22 | 3641 | BAE24765.1 | |
| AK143026.1 | 22 | 3708 | BAE25254.1 | |
| BC065137.1 | 20 | 2925 | AAH65137.1 | |
| X97817.1 | 21 | 3072 | CAA66397.1 |
More Information
Set Membership: |
|

Genomic Map