Assay Details
Target Gene Details
Entrez Gene ID: | 20524 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17 |
Gene Aliases: |
34kDa, 47kDa, PMP34, Pmp47 |
Location: |
Chr.15:81318921-81360765 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a17 | NM_011399.3 | 4 | 258 | NP_035529.1 |
AJ006341.1 | 4 | 270 | CAA06984.1 | |
AK002388.1 | 4 | 259 | BAB22062.1 | |
AK003672.1 | 4 | 245 | ||
BC008571.1 | 4 | 260 | AAH08571.1 | |
BC011292.1 | 4 | 240 | AAH11292.1 |
