Assay Details
Target Gene Details
Entrez Gene ID: | 16880 |
Gene Name: | leukemia inhibitory factor receptor |
Gene Aliases: |
A230075M04Rik, AW061234, LIF |
Location: |
Chr.15:7090611-7197489 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lifr | NM_001113386.1 | 6 | 869 | NP_001106857.1 |
| NM_013584.2 | 5 | 518 | NP_038612.1 | |
| AK145864.1 | 6 | 870 | BAE26706.1 | |
| AK155524.1 | 5 | 745 | BAE33308.1 | |
| BC031929.1 | 5 | 511 | AAH31929.1 | |
| BY716975.1 | ||||
| D17444.1 | 5 | 525 | BAA04258.1 | |
| D26177.1 | 5 | 525 | BAA05165.1 | |
| S73495.1 | 5 | 525 | AAC60697.1 | |
| S73496.1 | 5 | 525 | AAC60698.1 |
More Information
Set Membership: |
|

Genomic Map