Assay Details
Target Gene Details
Entrez Gene ID: | 239436 |
Gene Name: | solute carrier family 30 (zinc transporter), member 8 |
Gene Aliases: |
C820002P14Rik, ZnT-8, ZnT8 |
Location: |
Chr.15:52295553-52335733 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc30a8 | NM_172816.3 | NP_766404.1 | ||
| AK050482.1 | BAC34281.1 | |||
| AK050494.1 | BAC34289.1 | |||
| BC117544.1 | ||||
| BC118521.1 |
More Information
Set Membership: |
|

Genomic Map