Assay Details
Target Gene Details
Entrez Gene ID: | 74281 |
Gene Name: | spermatogenesis and centriole associated 1 |
Gene Aliases: |
1700084J23Rik, speriolin |
Location: |
Chr.15:76267835-76292577 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spatc1 | NM_028852.1 | NP_083128.1 | ||
| AB032199.1 | BAD08243.1 | |||
| AK006994.1 | BAB24825.1 | |||
| BC116725.1 | ||||
| BC118509.1 |

Genomic Map