Assay Details
Target Gene Details
Entrez Gene ID: | 20363 |
Gene Name: | selenoprotein P, plasma, 1 |
Gene Aliases: |
AU018766, D15Ucla1, Se-P, selp |
Location: |
Chr.15:3270767-3280508 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sepp1 | NM_001042613.1 | NP_001036078.1 | ||
NM_001042614.1 | NP_001036079.1 | |||
AI892015.1 | ||||
CK627189.1 |
