Assay Details
Target Gene Details
Entrez Gene ID: | 20363 |
Gene Name: | selenoprotein P, plasma, 1 |
Gene Aliases: |
AU018766, D15Ucla1, Se-P, selp |
Location: |
Chr.15:3270767-3280508 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sepp1 | NM_001042613.1 | NP_001036078.1 | ||
NM_001042614.1 | NP_001036079.1 | |||
NM_009155.3 | NP_033181.3 | |||
AI892015.1 | ||||
AK002450.1 | BAC55246.3 | |||
AK077265.1 | BAC55264.3 | |||
AK146774.1 | BAE27423.1 | |||
AK148623.1 | BAE28626.1 | |||
AK159269.1 | BAE34948.1 | |||
AK159524.1 | BAE35153.1 | |||
AK159894.1 | BAE35460.1 | |||
AK159911.1 | BAE35474.1 | |||
AK159935.1 | BAE35495.1 | |||
AK167611.1 | BAE39664.1 | |||
AK168463.1 | BAE40359.1 | |||
AK168583.1 | BAE40452.1 | |||
AK168631.1 | BAE40491.1 | |||
AK168747.1 | BAE40587.1 | |||
AK168926.1 | BAE40739.1 | |||
AK169011.1 | BAE40808.1 | |||
AK169158.1 | BAE40937.1 | |||
AK169362.1 | BAE41111.1 | |||
AK188386.1 | ||||
BC001991.1 | AAH01991.2 | |||
CB231357.1 | ||||
CK627189.1 |
