Assay Details
Target Gene Details
Entrez Gene ID: | 18414 |
Gene Name: | oncostatin M receptor |
Gene Aliases: |
OSMRB |
Location: |
Chr.15:6813568-6875246 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Osmr | NM_011019.3 | 8 | 1179 | NP_035149.2 |
| AB015978.1 | 8 | 1833 | BAA33725.1 | |
| AF058805.1 | 8 | 1149 | AAC40122.1 | |
| AK087179.1 | 8 | 1137 | ||
| AK157508.1 | 3 | 349 | ||
| BC137703.1 | 7 | 1074 |
More Information
Set Membership: |
|

Genomic Map