Assay Details
Target Gene Details
Entrez Gene ID: | 78757 |
Gene Name: | RPTOR independent companion of MTOR, complex 2 |
Gene Aliases: |
4921505C17Rik, 6030405M08Rik, AVO3, AW492497, D530039E11Rik |
Location: |
Chr.15:6708349-6800401 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rictor | NM_030168.3 | 12 | 1039 | NP_084444.3 |
AK036149.1 | 12 | 1040 | BAC29321.2 | |
AK082586.1 | 12 | 1030 | ||
AY497009.1 | 12 | 1017 | AAR89074.1 | |
AY540053.1 | 12 | 1037 | AAS46920.1 | |
BC022906.1 | 7 | 585 | AAH22906.1 | |
BC120903.1 | 8 | 678 | ||
BC120904.1 | 12 | 1037 |
