Assay Details
Target Gene Details
Entrez Gene ID: | 71474 |
Gene Name: | protein phosphatase 6, regulatory subunit 2 |
Gene Aliases: |
1110033O10Rik, 8430411H09Rik, B230107H12Rik, Pp6r2, Saps2 |
Location: |
Chr.15:89211521-89287015 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp6r2 | NM_026813.1 | 20 | 2540 | NP_081089.1 |
| AK173000.1 | 15 | 1723 | BAD32278.1 | |
| BC024879.1 | 19 | 2343 | AAH24879.1 | |
| BC112914.1 | 8 | 699 |
More Information
Set Membership: |
|

Genomic Map