Assay Details
Target Gene Details
Entrez Gene ID: | 406220 |
Gene Name: | keratin 77 |
Gene Aliases: |
4732484G22Rik, K1emb, Krt239, mK1b |
Location: |
Chr.15:101858731-101869706 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Krt77 | NM_001003667.1 | NP_001003667.1 | ||
AK132513.1 | ||||
AY379112.1 | AAR23930.1 | |||
BC107395.2 | ||||
BC107396.2 | ||||
BC141259.1 |
